siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 27, 28 | 5239 | ||
| GenBank mRNA | AF237953.1 | - | 25, 26 | 3256 | ||
| AK144114.1 | - | 3, 4 | 499 | |||
| AK220471.1 | - | 5, 6 | 1137 | |||
| BC038056.1 | - | 22, 23 | 3347 | |||
| BC054751.1 | - | 27, 28 | 4518 | |||
| BC055289.1 | - | 28, 29 | 5042 | |||
| BC096538.1 | - | 27, 28 | 5239 | |||

Pcnxl3
Genomic Map