siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290733.1 | NP_001277662.1 | 12 | 1697 | ||
| NM_145525.3 | NP_663500.2 | 11 | 1457 | |||
| GenBank mRNA | AK044411.1 | - | 12 | 1668 | ||
| AK220498.1 | - | 11 | 1404 | |||
| BC022908.1 | - | 6 | 616 | 5043261 | ||

Osbpl6
Genomic Map