siRNA Details
| Gene Symbol: Rell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145923.4 | NP_666035.2 | 4 | 581 | ||
| GenBank mRNA | AK133845.1 | - | 5 | 601 | ||
| AK140108.1 | - | 4 | 581 | |||
| AK159944.1 | - | 4 | 507 | |||
| AK161061.1 | - | 4 | 507 | |||
| AK182372.1 | - | 3 | 140 | |||
| AK202598.1 | - | 3 | 140 | |||
| BC024679.1 | - | 1 | 30 | |||
| BC031198.1 | - | 4 | 550 | 5059693 | ||
| BC066137.1 | - | 4 | 560 | 30012041 | ||
| BC066160.1 | - | 4 | 560 | 30012671 | ||

Rell1
Genomic Map