siRNA Details
| Gene Symbol: Zfp282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146175.3 | NP_666287.3 | 8 | 2568 | ||
| GenBank mRNA | AK030516.1 | - | 8 | 2476 | ||
| AK030919.1 | - | 1 | 1020 | |||
| AK088531.1 | - | 8 | 2584 | |||
| AK212078.1 | - | 1 | 36 | |||
| BC025581.1 | - | 9 | 2090 | 5325029 | ||

Zfp282
Genomic Map