siRNA Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146207.2 | NP_666319.2 | 8 | 854 | ||
| GenBank mRNA | AK144802.1 | - | 8 | 836 | ||
| AK150237.1 | - | 8 | 833 | |||
| AK170722.1 | - | 8 | 836 | |||
| AK196574.1 | - | 1 | 24 | |||
| AK201678.1 | - | 1 | 24 | |||
| BC010211.1 | - | 8 | 832 | 3969515 | ||
| BC024113.1 | - | 4 | 326 | 5097944 | ||
| BC027006.1 | - | 4 | 374 | |||

Cul4a
Genomic Map