siRNA Details
| Gene Symbol: Plekho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153119.3 | NP_694759.1 | 3 | 383 | ||
| GenBank mRNA | AK028069.1 | - | 3 | 374 | ||
| AK154092.1 | - | 3 | 278 | |||
| AK158288.1 | - | 3 | 351 | |||
| AK170479.1 | - | 3 | 367 | |||
| BC028908.1 | - | 3 | 342 | 3668775 | ||

Plekho2
Genomic Map