siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 11, 12 | 1667 | ||
| GenBank mRNA | AK035701.1 | - | 11, 12 | 1566 | ||
| AK098114.1 | - | 11, 12 | 1686 | |||
| AK146825.1 | - | 11, 12 | 1594 | |||
| BC023804.1 | - | 12, 13 | 1721 | 5352941 | ||
| BC078641.1 | - | 11, 12 | 2579 | |||

Dcaf8
Genomic Map