siRNA Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153570.2 | NP_705798.2 | 3 | 301 | ||
| GenBank mRNA | AK052601.1 | - | 3 | 301 | ||
| AK088355.1 | - | 3 | 302 | |||
| AK090024.1 | - | 3 | 314 | |||
| AK146006.1 | - | 3 | 297 | |||
| AK172310.1 | - | 3 | 280 | |||
| BC024616.1 | - | 3 | 291 | 4218310 | ||

Noc4l
Genomic Map