siRNA Details
| Gene Symbol: Cpne7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170684.2 | NP_733785.1 | 6 | 750 | ||
| GenBank mRNA | AK049823.1 | - | 6 | 679 | ||
| AK162900.1 | - | 6 | 751 | |||
| AU079878.1 | - | 6 | 669 | |||
| BC119328.1 | - | 6 | 649 | 8734077 | ||
| BC137595.1 | - | 6 | 649 | 8860614 | ||

Cpne7
Genomic Map