siRNA Details
| Gene Symbol: Slc24a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110240.1 | NP_001103710.1 | 1 | 973 | ||
| NM_172426.2 | NP_766014.1 | 2 | 1198 | |||
| GenBank mRNA | AK034424.1 | - | 2 | 1082 | ||
| AK038634.1 | - | 2 | 1082 | |||
| AK038800.1 | - | 2 | 1126 | |||
| AK083127.1 | - | 2 | 1082 | |||
| AK136058.1 | - | 2 | 1127 | |||
| BC115867.1 | - | 1 | 975 | 40081382 | ||
| BC116637.1 | - | 1 | 975 | 40081381 | ||

Slc24a2
Genomic Map