siRNA Details
| Gene Symbol: Gabrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008070.3 | NP_032096.1 | 10 | 7236 | ||
| GenBank mRNA | AK021039.1 | - | 1 | 802 | ||
| AK032113.1 | - | 1 | 2071 | |||
| AK046796.1 | - | 1 | 1851 | |||
| AK080856.1 | - | 1 | 478 | |||
| BC079663.1 | - | 1 | 714 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_172432.1 | - | 802 | Chr. - - [-] on Build | ||

Gabrb2
Genomic Map