siRNA Details
| Gene Symbol: D730001G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021322.1 | - | 2 | 631 | ||
| AK037167.1 | - | 2 | 630 | |||
| BC055822.1 | - | 3 | 694 | |||
| BC147771.1 | - | 1 | 510 | |||
| BC147774.1 | - | 1 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730001G18Rik| Gene Symbol: D730001G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021322.1 | - | 2 | 631 | ||
| AK037167.1 | - | 2 | 630 | |||
| BC055822.1 | - | 3 | 694 | |||
| BC147771.1 | - | 1 | 510 | |||
| BC147774.1 | - | 1 | 510 | |||