siRNA Details
| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012518.3 | NP_001012536.2 | 16 | 2488 | ||
| NM_001109686.2 | NP_001103156.1 | 15 | 2329 | |||
| NM_001109687.2 | NP_001103157.1 | 15 | 2344 | |||
| NM_172545.4 | NP_766133.2 | 16 | 2467 | |||
| GenBank mRNA | AB205007.1 | - | 16 | 2494 | ||
| AK049454.1 | - | 13 | 1738 | |||
| AK052174.1 | - | 16 | 2451 | |||
| AK082062.1 | - | 16 | 2478 | |||
| BC056938.1 | - | 13 | 2045 | |||
| BC089302.1 | - | 13 | 2197 | |||

Ehmt1
Genomic Map