siRNA Details
| Gene Symbol: Mzt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175245.4 | NP_780454.1 | 2 | 240 | ||
| GenBank mRNA | AK010788.1 | - | 2 | 216 | ||
| AK082817.1 | - | 2 | 186 | |||
| AK087564.1 | - | 2 | 184 | |||
| AK150268.1 | - | 2 | 216 | |||
| AK151232.1 | - | 2 | 216 | |||
| AK151358.1 | - | 2 | 216 | |||
| AK151819.1 | - | 2 | 216 | |||
| AK160911.1 | - | 2 | 225 | |||
| AK165605.1 | - | 2 | 203 | |||
| AK168114.1 | - | 2 | 181 | |||
| AK169036.1 | - | 2 | 190 | |||
| AK170746.1 | - | 2 | 190 | |||
| AK209030.1 | - | 1 | 66 | |||
| BB844588.1 | - | 2 | 241 | |||
| BC038168.1 | - | 2 | 178 | |||
| BC117005.1 | - | 2 | 215 | 40126324 | ||
| BC117007.1 | - | 2 | 215 | 40126326 | ||

Mzt1
Genomic Map