siRNA Details
| Gene Symbol: Sult2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199306.1 | NP_001186235.1 | 6 | 912 | ||
| NM_175250.5 | NP_780459.1 | 6 | 962 | |||
| GenBank mRNA | AK041070.1 | - | 6 | 962 | ||
| AK050422.1 | - | 6 | 910 | |||
| BC031851.1 | - | 6 | 858 | 4193548 | ||
| EU486166.1 | - | 6 | 962 | |||
| EU486167.1 | - | 5 | 864 | |||

Sult2a8
Genomic Map