siRNA Details
| Gene Symbol: Shisa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175259.4 | NP_780468.1 | 5 | 1180 | ||
| GenBank mRNA | AK134719.1 | - | 3 | 963 | ||
| AK192563.1 | - | 1 | 210 | |||
| AK212908.1 | - | 1 | 292 | |||
| BC044813.1 | - | 5 | 1182 | 4950177 | ||

Shisa4
Genomic Map