siRNA Details
| Gene Symbol: Notumos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028718.1 | - | 1 | 2316 | ||
| AK144387.1 | - | 1 | 1593 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Notumos| Gene Symbol: Notumos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028718.1 | - | 1 | 2316 | ||
| AK144387.1 | - | 1 | 1593 | |||