siRNA Details
| Gene Symbol: Bora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175265.4 | NP_780474.1 | 7, 8 | 829 | ||
| GenBank mRNA | AK034930.1 | - | 7, 8 | 830 | ||
| AK047285.1 | - | 1, 2 | 32 | |||
| AK141389.1 | - | 7, 8 | 780 | |||
| AK167430.1 | - | 7, 8 | 761 | |||
| BC052183.1 | - | 7, 8 | 730 | 6336038 | ||

Bora
Genomic Map