siRNA Details
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040152.1 | - | 6 | 2179 | ||
| BC150679.1 | - | 6 | 1882 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nav2| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040152.1 | - | 6 | 2179 | ||
| BC150679.1 | - | 6 | 1882 | |||