siRNA Details
| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175277.4 | NP_780486.1 | 4 | 377 | ||
| GenBank mRNA | AK019037.1 | - | 3 | 212 | ||
| AK028102.1 | - | 4 | 378 | |||
| AK131950.1 | - | 386 | ||||
| AK160488.1 | - | 4 | 381 | |||
| AK161878.1 | - | 149 | ||||
| AK218122.1 | - | 2 | 194 | |||
| BC049748.1 | - | 4 | 397 | |||
| BC116947.1 | - | 5 | 415 | 40126266 | ||
| BC116949.1 | - | 5 | 415 | 40126268 | ||

Bola3
Genomic Map