siRNA Details
| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175280.3 | NP_780489.2 | 7 | 1214 | ||
| GenBank mRNA | AK038772.1 | - | 7 | 1215 | ||
| AK086391.1 | - | 968 | ||||
| Gene Symbol: Gm6313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139668.1 | - | 1 | 56 | ||

Cfap61
Genomic Map