siRNA Details
| Gene Symbol: D930048N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086740.1 | - | 4 | 548 | ||
| BC075670.1 | - | 4 | 530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D930048N14Rik| Gene Symbol: D930048N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086740.1 | - | 4 | 548 | ||
| BC075670.1 | - | 4 | 530 | |||