siRNA Details
| Gene Symbol: Eftud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159672.1 | NP_001153144.1 | 17 | 2063 | ||
| NM_175317.3 | NP_780526.2 | 17 | 2092 | |||
| GenBank mRNA | AK028676.1 | - | 17 | 2065 | ||
| AK031647.1 | - | 17 | 2049 | |||
| BC031852.1 | - | 11 | 1338 | |||
| BC045616.1 | - | 17 | 2046 | 4983234 | ||

Eftud1
Genomic Map