siRNA Details
| Gene Symbol: D330045A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175326.5 | NP_780535.1 | 1 | 719 | ||
| GenBank mRNA | AK052376.1 | - | 1 | 643 | ||
| AK155588.1 | - | 1 | 716 | |||
| AK171459.1 | - | 1 | 718 | |||
| BC112416.1 | - | 1 | 619 | |||
| BC113128.1 | - | 1 | 619 | 40061080 | ||

D330045A20Rik
Genomic Map