siRNA Details
| Gene Symbol: E130012A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175332.3 | NP_780541.2 | 1 | 1145 | ||
| GenBank mRNA | AK087411.1 | - | 1 | 1132 | ||
| BC006054.1 | - | 1 | 584 | |||
| BC055770.1 | - | 1 | 1145 | 6403695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130012A19Rik| Gene Symbol: E130012A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175332.3 | NP_780541.2 | 1 | 1145 | ||
| GenBank mRNA | AK087411.1 | - | 1 | 1132 | ||
| BC006054.1 | - | 1 | 584 | |||
| BC055770.1 | - | 1 | 1145 | 6403695 | ||