siRNA Details
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006270.4 | NP_006261.1 | 6 | 715 | ||
GenBank mRNA | BC016286.1 | - | 6 | 694 | 3910343 | |
BC016318.1 | - | 6 | 719 | 4073829 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006270.4 | NP_006261.1 | 6 | 715 | ||
GenBank mRNA | BC016286.1 | - | 6 | 694 | 3910343 | |
BC016318.1 | - | 6 | 719 | 4073829 |