siRNA Details
| Gene Symbol: Ncapd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178113.3 | NP_835214.2 | 3 | 599 | ||
| GenBank mRNA | AK014555.1 | - | 2 | 388 | ||
| AK019297.1 | - | 2 | 168 | |||
| AK129046.1 | - | 2 | 125 | |||
| AK139081.1 | - | 2 | 157 | |||
| BC048190.1 | - | 3 | 599 | 6831876 | ||
| DQ340802.1 | - | 2 | 254 | |||

Ncapd3
Genomic Map