siRNA Details
| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178406.2 | NP_848493.1 | 2 | 393 | ||
| GenBank mRNA | BC029098.1 | - | 1 | 299 | 3481871 | |
| BC037650.1 | - | 1 | 392 | 5151632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr153| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178406.2 | NP_848493.1 | 2 | 393 | ||
| GenBank mRNA | BC029098.1 | - | 1 | 299 | 3481871 | |
| BC037650.1 | - | 1 | 392 | 5151632 | ||