siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163328.1 | NP_001156800.1 | 6 | 774 | ||
| NM_001163329.1 | NP_001156801.1 | 5 | 367 | |||
| NM_001163330.1 | NP_001156802.1 | 5 | 367 | |||
| NM_178631.4 | NP_848746.1 | 7 | 867 | |||
| GenBank mRNA | AK032086.1 | - | 6 | 707 | ||
| AK038465.1 | - | 5 | 345 | |||
| AK079659.1 | - | 6 | 1596 | |||
| BC052358.1 | - | 5 | 328 | 6534677 | ||
| BY252332.1 | - | 5 | 368 | |||

Ralyl
Genomic Map