siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163328.1 | NP_001156800.1 | 9 | 1317 | ||
| NM_001163329.1 | NP_001156801.1 | 8 | 910 | |||
| NM_001163330.1 | NP_001156802.1 | 7 | 737 | |||
| NM_178631.4 | NP_848746.1 | 10 | 1410 | |||
| GenBank mRNA | AK032086.1 | - | 9 | 1250 | ||
| AK038465.1 | - | 8 | 888 | |||
| AK079659.1 | - | 9 | 2139 | |||
| BC052358.1 | - | 8 | 871 | 6534677 | ||

Ralyl
Genomic Map