siRNA Details
| Gene Symbol: Klhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178633.3 | NP_848748.2 | 597 | |||
| GenBank mRNA | AK032116.1 | - | 145 | |||
| AK167903.1 | - | 388 | ||||
| AK220570.1 | - | 597 | ||||
| BC031142.1 | - | 468 | 4972965 | |||
| BC031144.1 | - | 491 | 4973325 | |||

Klhl2
Genomic Map