siRNA Details
| Gene Symbol: Sfxn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178639.4 | NP_848754.1 | 1, 2 | 120 | ||
| GenBank mRNA | AF325264.1 | - | 1, 2 | 138 | ||
| AK036285.1 | - | 1, 2 | 118 | |||
| AK043706.1 | - | 1, 2 | 119 | |||
| AK082073.1 | - | 1, 2 | 118 | |||
| BB628882.1 | - | 1, 2 | 121 | |||
| BC089588.1 | - | 1, 2 | 120 | |||

Sfxn5
Genomic Map