siRNA Details
| Gene Symbol: ITSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006277.2 | NP_006268.2 | 3 | 349 | ||
| NM_019595.3 | NP_062541.3 | 3 | 349 | |||
| NM_147152.2 | NP_671494.2 | 3 | 349 | |||
| GenBank mRNA | AB033082.1 | - | 3 | 256 | ||
| AF182198.1 | - | 3 | 328 | |||
| AF182199.1 | - | 3 | 328 | |||
| AF248540.1 | - | 2 | 130 | |||
| BC020921.2 | - | 2 | 126 | |||
| BC038963.1 | - | 3 | 340 | |||
| BC146779.1 | - | 3 | 256 | 9007231 | ||
| DA382952.1 | - | 3 | 599 | |||

ITSN2
Genomic Map