siRNA Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178642.5 | NP_848757.4 | 14, 15 | 1621 | ||
| GenBank mRNA | AK032793.1 | - | 9, 10 | 633 | ||
| AK052589.1 | - | 14, 15 | 1573 | |||
| BC004697.1 | - | 6, 7 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano1| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178642.5 | NP_848757.4 | 14, 15 | 1621 | ||
| GenBank mRNA | AK032793.1 | - | 9, 10 | 633 | ||
| AK052589.1 | - | 14, 15 | 1573 | |||
| BC004697.1 | - | 6, 7 | 487 | |||