siRNA Details
| Gene Symbol: Oaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178644.3 | NP_848759.1 | 4 | 1871 | ||
| GenBank mRNA | AK051349.1 | - | 4 | 1871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oaf| Gene Symbol: Oaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178644.3 | NP_848759.1 | 4 | 1871 | ||
| GenBank mRNA | AK051349.1 | - | 4 | 1871 | ||