siRNA Details
| Gene Symbol: Dcun1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190733.1 | NP_001177662.1 | 9 | 820 | ||
| NM_001190734.1 | NP_001177663.1 | 11 | 860 | |||
| NM_178896.5 | NP_849227.1 | 10 | 925 | |||
| GenBank mRNA | AK033559.1 | - | 10 | 870 | ||
| AK076991.1 | - | 11 | 861 | |||
| BC031793.1 | - | 1 | 49 | |||
| BC060213.1 | - | 10 | 867 | |||
| BC108346.1 | - | 10 | 827 | 6310392 | ||
| BC137627.1 | - | 10 | 869 | 8860646 | ||
| BC137628.1 | - | 10 | 869 | 8860647 | ||
| BC144794.1 | - | 9 | 764 | 9053342 | ||
| CO433728.1 | - | 5 | 429 | |||

Dcun1d4
Genomic Map