siRNA Details
| Gene Symbol: Wdr92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178909.4 | NP_849240.1 | 4 | 529 | ||
| GenBank mRNA | AK033294.1 | - | 4 | 475 | ||
| AK047691.1 | - | 4 | 504 | |||
| AK088218.1 | - | 4 | 504 | |||
| AK141504.1 | - | 1 | 1282 | |||
| AK142228.1 | - | 4 | 475 | |||
| BC096540.1 | - | 4 | 458 | 6403681 | ||

Wdr92
Genomic Map