siRNA Details
| Gene Symbol: Wdr92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178909.4 | NP_849240.1 | 8 | 1816 | ||
| GenBank mRNA | AK033294.1 | - | 8 | 1762 | ||
| AK088218.1 | - | 8 | 1791 | |||
| AK142228.1 | - | 8 | 1763 | |||
| BC096540.1 | - | 8 | 1746 | 6403681 | ||

Wdr92
Genomic Map