siRNA Details
| Gene Symbol: B230219D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181278.2 | NP_851795.1 | 2 | 3087 | ||
| GenBank mRNA | BC006718.1 | - | 1 | 335 | ||
| BC051430.1 | - | 2 | 3087 | 6530869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230219D22Rik| Gene Symbol: B230219D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181278.2 | NP_851795.1 | 2 | 3087 | ||
| GenBank mRNA | BC006718.1 | - | 1 | 335 | ||
| BC051430.1 | - | 2 | 3087 | 6530869 | ||