siRNA Details
| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181854.2 | NP_862902.1 | 2 | 437 | ||
| GenBank mRNA | AK129444.1 | - | 2 | 337 | ||
| AK132164.1 | - | 2 | 400 | |||
| AK133990.1 | - | 2 | 363 | |||
| AK139963.1 | - | 2 | 402 | |||
| AK170835.1 | - | 2 | 380 | |||
| BC028991.1 | - | 2 | 349 | 5355869 | ||

Champ1
Genomic Map