siRNA Details
| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181854.2 | NP_862902.1 | 2 | 1926 | ||
| GenBank mRNA | AK129444.1 | - | 2 | 1826 | ||
| AK132164.1 | - | 2 | 1889 | |||
| AK133990.1 | - | 2 | 1852 | |||
| AK139963.1 | - | 2 | 1892 | |||
| AK170835.1 | - | 2 | 1869 | |||
| BC028991.1 | - | 2 | 1838 | 5355869 | ||

Champ1
Genomic Map