siRNA Details
Gene Symbol: Ccp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182995.1 | NP_892040.1 | 4 | 532 | ||
GenBank mRNA | AK141641.1 | - | 4 | 527 | ||
BC053103.1 | - | 4 | 532 | 6406475 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182995.1 | NP_892040.1 | 4 | 532 | ||
GenBank mRNA | AK141641.1 | - | 4 | 527 | ||
BC053103.1 | - | 4 | 532 | 6406475 |