siRNA Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040686.1 | NP_001035776.1 | 5, 6 | 729 | ||
| NM_182996.3 | NP_892041.2 | 5, 6 | 791 | |||
| GenBank mRNA | AK154896.1 | - | 733 | |||
| AK170229.1 | - | 5, 6 | 794 | |||
| AK170358.1 | - | 4, 5 | 629 | |||
| BC044879.1 | - | 1, 2 | 62 | |||
| BC055041.1 | - | 5, 6 | 757 | |||
| BC145470.1 | - | 5, 6 | 688 | 9054032 | ||
| BC150776.1 | - | 5, 6 | 688 | 9087687 | ||

Zfp692
Genomic Map