siRNA Details
| Gene Symbol: Spinkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183123.2 | NP_898946.1 | 5 | 381 | ||
| GenBank mRNA | AK020535.1 | - | 5 | 382 | ||
| EU234015.1 | - | 758 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spinkl| Gene Symbol: Spinkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183123.2 | NP_898946.1 | 5 | 381 | ||
| GenBank mRNA | AK020535.1 | - | 5 | 382 | ||
| EU234015.1 | - | 758 | ||||