siRNA Details
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034891.3 | NP_001030063.1 | 5 | 737 | ||
| NM_001039552.2 | NP_001034641.2 | 1670 | ||||
| GenBank mRNA | AK019108.1 | - | 6 | 824 | ||
| AK160176.1 | - | 5 | 738 | |||
| BC150877.1 | - | 1731 | 9087788 | |||

Ermard
Genomic Map