siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183137.2 | NP_898960.2 | 10 | 999 | ||
| GenBank mRNA | AK019105.1 | - | 10 | 970 | ||
| AK154660.1 | - | 10 | 964 | |||
| AK196075.1 | - | 2 | 65 | |||
| BC049110.1 | - | 10 | 935 | |||
| BC064313.1 | - | 10 | 935 | |||
| BC094318.1 | - | 9 | 1488 | |||
| BC137580.1 | - | 10 | 968 | 8860599 | ||
| BC144724.1 | - | 10 | 968 | 9053272 | ||

Enthd2
Genomic Map