siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183137.2 | NP_898960.2 | 8 | 664 | ||
| GenBank mRNA | AK019105.1 | - | 8 | 635 | ||
| AK154660.1 | - | 8 | 629 | |||
| AK179031.1 | - | 1 | 43 | |||
| AK194765.1 | - | 1 | 43 | |||
| BC049110.1 | - | 8 | 600 | |||
| BC064313.1 | - | 8 | 600 | |||
| BC094318.1 | - | 7 | 1153 | |||
| BC137580.1 | - | 8 | 633 | 8860599 | ||
| BC144724.1 | - | 8 | 633 | 9053272 | ||

Enthd2
Genomic Map