siRNA Details
| Gene Symbol: Exoc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198103.2 | NP_932771.1 | 1 | 4095 | ||
| GenBank mRNA | AK054047.1 | - | 1 | 1181 | ||
| AK076456.1 | - | 1 | 358 | |||
| AK154344.1 | - | 1 | 980 | |||
| BC034060.1 | - | 1 | 2623 | |||
| BC057052.1 | - | 1 | 4095 | 6822067 | ||

Exoc8
Genomic Map