siRNA Details
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198297.3 | NP_938039.2 | 2, 3 | 159 | ||
| GenBank mRNA | AJ297969.1 | - | 2, 3 | 177 | ||
| AK030807.1 | - | 2, 3 | 342 | |||
| AK138735.1 | - | 225 | ||||
| BC104351.1 | - | 2, 3 | 159 | 40049558 | ||

Trat1
Genomic Map