siRNA Details
| Gene Symbol: HSPB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006308.2 | NP_006299.1 | 1 | 283 | ||
| GenBank mRNA | AK314215.1 | - | 1 | 284 | ||
| BC126272.1 | - | 1 | 138 | 8991988 | ||
| BC132869.1 | - | 1 | 150 | 40146891 | ||
| BX106905.1 | - | 1 | 268 | |||
| CR450314.1 | - | 1 | 94 | |||
| U15590.1 | - | 1 | 269 | |||
| Y17782.1 | - | 1 | 242 | |||

HSPB3
Genomic Map